(4-Ethoxy-3,5-difluorophenyl)acetic acid 97%

Supplier: Thermo Scientific
Warning

H31873.MDEA 63.8 EUR
H31873.MD H31873.03
(4-Ethoxy-3,5-difluorophenyl)acetic acid 97%
(4-Ethoxy-3,5-difluorophenyl)acetic acid
Formula: C₁₀H₁₀F₂O₃
MW: 216,18
Melting Pt: 55…58 °C
Storage Temperature: Ambient
MDL Number: MFCD09258681
CAS Number: 1017779-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR