Dimethylzinc ≥96%
Supplier: Thermo Scientific
|
Danger
|
39563.14EA
1130
EUR
39563.14
Dimethylzinc ≥96%
Dimethylzinc
Formula:
C₂H₆Zn MW: 95,478 g/mol Boiling Pt: 46 °C Melting Pt: –40 °C Density: 1,386 Flash Pt: –18 °C (–0,4 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014854 CAS Number: 544-97-8 EINECS: 208-884-1 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...