5-Chlorobenzoxazole 95%
Supplier: Thermo Scientific
|
Warning
|
H27083.06EA
79.9
EUR
H27083.06
5-Chlorobenzoxazole 95%
5-Chlorobenzoxazole
Formula:
C₇H₄ClNO MW: 153,57 g/mol Melting Pt: 39…41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216939 CAS Number: 17200-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...