6-Chloro-2-fluoro-3-methylbenzaldehyde ≥97%

Supplier: Thermo Scientific
Warning

B22937.03EA 34.4 EUR
B22937.03 B22937.06
6-Chloro-2-fluoro-3-methylbenzaldehyde ≥97%
6-Chloro-2-fluoro-3-methylbenzaldehyde
Formula: C₈H₆ClFO
MW: 172,59
Boiling Pt: 249 °C
Density: 1,300
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01631370
CAS Number: 286474-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR