5-Methylbenzoxazole ≥99%

Supplier: Thermo Scientific
Danger

B22596.14EA 63.4 EUR
B22596.14 B22596.06 B22596.03
5-Methylbenzoxazole ≥99%
5-Methylbenzoxazole
Formula: C₈H₇NO
MW: 133,15
Melting Pt: 41…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00216938
CAS Number: 10531-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR