5-Methylbenzoxazole

Supplier: Molekula

Synonyms: 5-Methylbenzo[d]oxazole

MOLEM54638396EA 0 EUR
MOLEM54638396
5-Methylbenzoxazole
5-Methylbenzoxazole
Formula: C₈H₇NO
MW: 133,15 g/mol
MDL Number: MFCD00216938
CAS Number: 10531-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR