3-Methoxy-2-(trifluoromethyl)benzonitrile ≥97%
|
Danger
|
H33717.03EA
93.2
EUR
H33717.03
H33717.MD
3-Methoxy-2-(trifluoromethyl)benzonitrile ≥97%
3-Methoxy-2-(trifluoromethyl)benzonitrile
Formula:
C₉H₆F₃NO MW: 201,15 g/mol Melting Pt: 103…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD13185684 CAS Number: 1214385-02-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...