2-Chloro-3-fluorobenzaldehyde 95%

Supplier: Thermo Scientific
Warning

H34238.03EA 121 EUR
H34238.03 H34238.MD
2-Chloro-3-fluorobenzaldehyde 95%
2-Chloro-3-fluorobenzaldehyde
Formula: C₇H₄ClFO
Melting Pt: 29…32 °C
Storage Temperature: Ambient
MDL Number: MFCD07782053
CAS Number: 96516-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR