Insulated shipper
Supplier: Sonoco
216-1339EA 481 EUR
216-1339
Insulated shipper
Mailers and Shipping Containers
EPS (expanded polystyrene) and Neopor.
  • Square format
  • Wall thickness: 50 mm
  • Reliable, lightweight and protective
These insulated containers are suitable for different packaging situations in pharmacies, trade and the food industry. They ensure excellent protection from heat and cold during the transport and despatch of medicines, food and other temperature-sensitive products.