Power supply, PS600
Supplier: Hoefer
732-2381EA 1980 EUR
HOEFPS600
Power supply, PS600
The midrange PS600 power supply is designed for electrophoresis and blotting techniques, including large format and high throughput applications. The small footprint, large handle, and simple operation make the PS600 easy to set up and use. With the large, easy to read LED, simply set the constant parameter, set the timer (if desired), and push start.
  • Constant voltage or constant current mode with automatic crossover
  • Four sets of four mm output jacks
  • Continuous runs or 999 minute timer
  • Automatic recovery after power failure
  • Open circuit detection
  • Short circuit protection