5-(2-Pyridyl)-1H-tetrazole

Supplier: Molekula

Synonyms: 2-(1H-Tetrazol-5-yl)pyridine5-(2-Pyridyl)-1H-tetrazole

MOLE73645424-1GEA 0 EUR
MOLE73645424-1G
5-(2-Pyridyl)-1H-tetrazole
5-(2-Pyridyl)-1H-tetrazole
Formula: C₆H₅N₅
MW: 147,14 g/mol
MDL Number: MFCD00068114
CAS Number: 33893-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR