5-(3-Pyridyl)-1H-tetrazole

Supplier: Molekula

Synonyms: 3-(2H-Tetrazol-5-yl)pyridine

MOLE58528284-1GEA 0 EUR
MOLE58528284-1G
5-(3-Pyridyl)-1H-tetrazole
5-(3-Pyridyl)-1H-tetrazole
Formula: C₆H₅N₅
MW: 147,14 g/mol
MDL Number: MFCD00040281
CAS Number: 3250-74-6
EINECS: 426-810-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR