4-[2-(Trifluoromethyl)phenoxy]piperidine ≥99%

Supplier: Thermo Scientific
Warning

H50813.03EA 317 EUR
H50813.03 H50813.MD
4-[2-(Trifluoromethyl)phenoxy]piperidine ≥99%
4-[2-(Trifluoromethyl)phenoxy]piperidine
Formula: C₁₂H₁₄F₃NO
MW: 245,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06248881
CAS Number: 824390-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR