4-(2-Methylphenoxy)piperidine ≥97%

Supplier: Thermo Scientific
Warning

H50951.MDEA 75.4 EUR
H50951.MD H50951.03
4-(2-Methylphenoxy)piperidine ≥97%
4-(2-Methylphenoxy)piperidine
Formula: C₁₂H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD06248344
CAS Number: 63843-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR