3-(2-Methylphenoxy)piperidine hydrochloride ≥98%

Supplier: Thermo Scientific
Warning

H59396.MDEA 72.2 EUR
H59396.MD
3-(2-Methylphenoxy)piperidine hydrochloride ≥98%
3-(2-Methylphenoxy)piperidine hydrochloride
Formula: C₁₂H₁₈ClNO
MW: 227,73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21606599
CAS Number: 1858256-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR