(Dimethylamino)acetonitrile 98%
Supplier: Thermo Scientific
|
Danger
|
B25085.22EA
99.5
EUR
B25085.22
B25085.14
(Dimethylamino)acetonitrile 98%
(Dimethylamino)acetonitrile
Formula:
C₄H₈N₂ MW: 84,12 g/mol Boiling Pt: 137…138 °C Density: 0,863 Flash Pt: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001890 CAS Number: 926-64-7 EINECS: 213-140-4 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...