1-Aminocyclohexanecarboxylic acid hydrochloride ≥98%

Supplier: Thermo Scientific
Warning

B24031.14EA 22.6 EUR
B24031.14 B24031.22
1-Aminocyclohexanecarboxylic acid hydrochloride ≥98%
1-Aminocyclohexanecarboxylic acid hydrochloride
Formula: C₇H₁₄ClNO₂
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00044481
CAS Number: 39692-17-6
EINECS: 254-594-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR