2-Mercapto-1,3,4-thiadiazole

Supplier: Molekula

Synonyms: 1,3,4-Thiadiazole-2-thiol

MOLE51938931-25GEA 0 EUR
MOLE51938931-25G
2-Mercapto-1,3,4-thiadiazole
2-Mercapto-1,3,4-thiadiazole
Formula: C₂H₂N₂S₂
MW: 118,17
MDL Number: MFCD00040404
CAS Number: 18686-82-3
EINECS: 242-504-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR