1H,1H,5H-Octafluoropentyl acrylate ≥97% stabilised

Supplier: Thermo Scientific
Warning

Synonyms: 2,2,3,3,4,4,5,5-Octafluoropentyl acrylate

Stabilisation: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)

L16561.14EA 145 EUR
L16561.14
1H,1H,5H-Octafluoropentyl acrylate ≥97% stabilised
1H,1H,5H-Octafluoropentyl acrylate
Formula: C₈H₆F₈O₂
MW: 286,12
Boiling Pt: 56 °C (8 mmHg)
Density: 1,481
Flash Pt: 71 °C (159 °F)
MDL Number: MFCD00039279
CAS Number: 376-84-1
EINECS: 206-816-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR