2-Ethoxy-1-naphthaldehyde ≥98%

Supplier: Thermo Scientific
Warning

B25306.03EA 27.9 EUR
B25306.03 B25306.06
2-Ethoxy-1-naphthaldehyde ≥98%
2-Ethoxy-1-naphthaldehyde
Formula: C₁₃H₁₂O₂
Melting Pt: 111…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00046427
CAS Number: 19523-57-0
EINECS: 243-126-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR