3-Phenylphthalide ≥99%

Synonyms: 3-Phenylisobenzofuran-1(3H)-one

L16181.14EA 192 EUR
L16181.14 L16181.06
3-Phenylphthalide ≥99%
3-Phenylphthalide
Formula: C₁₄H₁₀O₂
MW: 210,23 g/mol
Melting Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00023099
CAS Number: 5398-11-8
EINECS: 226-426-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR