(4-Methylphenylthio)acetone ≥97%

Supplier: Thermo Scientific

B20247.06EA 44.6 EUR
B20247.06 B20247.03
(4-Methylphenylthio)acetone ≥97%
(4-Methylphenylthio)acetone
Formula: C₁₀H₁₂OS
MW: 180,27
Boiling Pt: 96…98 °C (0,1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00052713
CAS Number: 1200-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR