Overalls with bib and braces for men, C.I., design c, dark grey

Supplier: Fristads Kansas Group
KWIN870941210C52EA 0 EUR
KWIN870941210C52 KWIN870941131C44 KWIN870941131C156 KWIN870941941C50 KWIN870941520C54 KWIN870941530C154 KWIN870941940C54 KWIN870941941C44 KWIN870941940C48 KWIN870941941C150 KWIN870941331C58 KWIN870941131C62 KWIN870941941C54 KWIN870941940C46 KWIN870941331C46 KWIN870941331C48 KWIN870941131C150 KWIN870941131C48 KWIN870941231C44 KWIN870941640C48 KWIN870941210C154 KWIN870941440C50 KWIN870941210C56 KWIN870941539C60 KWIN870941231C46 KWIN870941210C148 KWIN870941941C56 KWIN870941640C58 KWIN870941210C146 KWIN870941941C60 KWIN870941941C156 KWIN870941440C148 KWIN870941210C150 KWIN870941331C152 KWIN870941440C150 KWIN870941539C62 KWIN870941940C148 KWIN870941640C62 KWIN870941210C44 KWIN870941231C50 KWIN870941539C52 KWIN870941440C60 KWIN870941530C52 KWIN870941730C154 KWIN870941530C156 KWIN870941520C46 KWIN870941131C146 KWIN870941941C146 KWIN870941940C150 KWIN870941640C156 KWIN870941231C48 KWIN870941539C148 KWIN870941520C146 KWIN870941539C56 KWIN870941940C52 KWIN870941640C54 KWIN870941530C44 KWIN870941520C58 KWIN870941331C62 KWIN870941210C156 KWIN870941539C46 KWIN870941539C54 KWIN870941131C152 KWIN870941331C52 KWIN870941941C52 KWIN870941940C156 KWIN870941520C156 KWIN870941331C50 KWIN870941520C62 KWIN870941440C154 KWIN870941730C62 KWIN870941730C56 KWIN870941530C50 KWIN870941539C44 KWIN870941730C50 KWIN870941231C156 KWIN870941640C152 KWIN870941640C56 KWIN870941440C58 KWIN870941530C62 KWIN870941131C46 KWIN870941539C156 KWIN870941940C152 KWIN870941440C62 KWIN870941131C60 KWIN870941941C46 KWIN870941530C146 KWIN870941941C152 KWIN870941640C148 KWIN870941520C154 KWIN870941210C62 KWIN870941940C56 KWIN870941331C44 KWIN870941231C58 KWIN870941730C48 KWIN870941730C150 KWIN870941231C52 KWIN870941231C150 KWIN870941440C146 KWIN870941520C148 KWIN870941520C48 KWIN870941131C58 KWIN870941440C46 KWIN870941331C148 KWIN870941941C148 KWIN870941520C56 KWIN870941940C44 KWIN870941231C152 KWIN870941941C62 KWIN870941730C44 KWIN870941231C56 KWIN870941131C56 KWIN870941131C52 KWIN870941730C152 KWIN870941530C148 KWIN870941940C50 KWIN870941730C156 KWIN870941520C44 KWIN870941210C48 KWIN870941539C48 KWIN870941539C50 KWIN870941231C148 KWIN870941331C154 KWIN870941640C50 KWIN870941331C156 KWIN870941530C150 KWIN870941940C146 KWIN870941440C56 KWIN870941331C60 KWIN870941440C52 KWIN870941331C54 KWIN870941539C58 KWIN870941520C152 KWIN870941941C58 KWIN870941640C44 KWIN870941440C48 KWIN870941331C146 KWIN870941331C56 KWIN870941640C150 KWIN870941530C58 KWIN870941941C48 KWIN870941131C54 KWIN870941730C146 KWIN870941640C52 KWIN870941440C54 KWIN870941530C56 KWIN870941210C60 KWIN870941231C146 KWIN870941530C48 KWIN870941539C146 KWIN870941539C150 KWIN870941940C62 KWIN870941640C60 KWIN870941210C46 KWIN870941131C148 KWIN870941520C50 KWIN870941331C150 KWIN870941530C46 KWIN870941131C154 KWIN870941131C50 KWIN870941640C146 KWIN870941210C58 KWIN870941940C60 KWIN870941539C152 KWIN870941520C60 KWIN870941231C54 KWIN870941520C52 KWIN870941530C60 KWIN870941539C154 KWIN870941210C50 KWIN870941210C54 KWIN870941440C152 KWIN870941730C58 KWIN870941231C62 KWIN870941730C52 KWIN870941640C154 KWIN870941530C54 KWIN870941940C154 KWIN870941440C156 KWIN870941231C154 KWIN870941730C46 KWIN870941520C150 KWIN870941440C44 KWIN870941730C148 KWIN870941730C60 KWIN870941640C46 KWIN870941940C58 KWIN870941530C152 KWIN870941730C54 KWIN870941231C60 KWIN870941210C152 KWIN870941941C154
Overalls with bib and braces for men, C.I., design c, dark grey
Workwear
These overalls with bib and braces are made from 65% polyester, 35% cotton, with a soft nap on reverse side.

  • Fly with zip
  • Two chest pockets, one with flap, the other with pen pocket
  • Two front pockets, two back pockets with flap
  • Leg pocket with flap and three small pockets
  • Folding-rule pocket, leg pocket with mobile phone pocket with flap and two small pockets
  • Adjustable waist, side openings
  • Knee pockets that open from the inside and can accommodate height-adjustable kneepads

Weight: 260 g/m²

Basic colour: Dark grey

Ordering information: Available with 12 different accent colours.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR