Rotary piston tubing pump drive
Supplier: Ismatec
ISMCISM919AEA 4290 EUR
ISMCISM919A
Rotary piston tubing pump drive
The ideal pump system is for corrosive media and very accurate dispensing. The dispensing pump can be calibrated, it contains memory enough to store four sets of operating parameters.
  • Process suitable and programmable piston pump
  • Dispensing and delivery pump
  • Dust-tight and waterproof
  • Washdown pump with stainless steel housing
  • RS232 and analog interface