4-Carboxy-2-fluorophenylboronic acid pinacol ester 96%
|
Warning
|
Synonyms:
4-Carboxy-2-fluorophenylboronic Acid Pinacol Ester
, 4-Carboxy-2-fluorobenzeneboronic acid pinacol ester
H62028.MDEA
29.5
EUR
H62028.MD
4-Carboxy-2-fluorophenylboronic acid pinacol ester 96%
4-Carboxy-2-fluorophenylboronic acid pinacol ester
Formula:
C₁₃H₁₆BFO₄ MW: 266,07 g/mol Melting Pt: 199…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD12756466 CAS Number: 1050423-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...