2-Cyano-3-fluorophenylboronic acid pinacol ester ≥96%

Synonyms: 2-Cyano-3-fluorobenzeneboronic acid pinacol ester2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

H62371.03EA 430 EUR
H62371.03 H62371.MD
2-Cyano-3-fluorophenylboronic acid pinacol ester ≥96%
2-Cyano-3-fluorophenylboronic acid pinacol ester
Formula: C₁₃H₁₅BFNO₂
MW: 247,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878541
CAS Number: 765916-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR