4-(2-Fluoro-4-nitrophenyl)morpholine ≥98%

Supplier: Thermo Scientific
Warning

H61940.06EA 19 EUR
H61940.06 H61940.14 H61940.22
4-(2-Fluoro-4-nitrophenyl)morpholine ≥98%
4-(2-Fluoro-4-nitrophenyl)morpholine
Formula: C₁₀H₁₁FN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD03138381
CAS Number: 2689-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR