4-(Isopropoxycarbonyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(Isopropoxycarbonyl)benzeneboronic acid(4-(Isopropoxycarbonyl)phenyl)boronic acid

H53335.06EA 440 EUR
H53335.06
4-(Isopropoxycarbonyl)phenylboronic acid ≥98%
4-(Isopropoxycarbonyl)phenylboronic acid
Formula: C₁₀H₁₃BO₄
MW: 208,02 g/mol
Melting Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD02259385
CAS Number: 342002-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR