2-Benzyloxy-3-fluorophenylboronic acid pinacol ester ≥96%

Supplier: Thermo Scientific

Synonyms: 2-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester

H62186.MDEA 87.8 EUR
H62186.MD H62186.03
2-Benzyloxy-3-fluorophenylboronic acid pinacol ester ≥96%
2-Benzyloxy-3-fluorophenylboronic acid pinacol ester
Formula: C₁₉H₂₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD23098817
CAS Number: 1661043-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR