4-(1H-Pyrazol-1-yl)benzoic acid ≥90%, tech.
Supplier: Thermo Scientific
|
Warning
|
H56405.06EA
490
EUR
H56405.06
H56405.03
4-(1H-Pyrazol-1-yl)benzoic acid ≥90%, tech.
4-(1H-Pyrazol-1-yl)benzoic acid
Formula:
C₁₀H₈N₂O₂ MW: 188,18 g/mol Melting Pt: 266 °C Storage Temperature: Ambient |
MDL Number:
MFCD02682056 CAS Number: 16209-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...