2-(1-Naphthylmethyl)-2-imidazoline hydrochloride

Supplier: EDQM

Synonyms: AlbalonNaphazoline Hydrochloride

EDQMN0080000EA 192 EUR
EDQMN0080000
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride
Formula: C₁₄H₁₅ClN₂
MW: 246,738 g/mol
MDL Number: MFCD00012554
CAS Number: 550-99-2
EINECS: 208-989-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR