2-(Tributylstannyl)benzo[b]thiophene 95%
Supplier: Thermo Scientific
|
Danger
|
H55348.MDEA
49
EUR
H55348.MD
2-(Tributylstannyl)benzo[b]thiophene 95%
2-(Tributylstannyl)benzo[b]thiophene
Formula:
C₂₀H₃₂SSn MW: 423,22 g/mol Density: 1,184 Storage Temperature: Ambient |
MDL Number:
MFCD01318936 CAS Number: 148961-88-0 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...