5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate
Supplier: Molekula
Synonyms:
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4''-disulphonic acid monosodium salt hydrate
, PDT-sulphonate monosodium salt hydrate
, FerroZine iron reagent hydrate
, PDT disulphonate monosodium salt hydrate
MOLEM16507877EA
0
EUR
MOLEM16507877
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate
Formula:
C₂₀H₁₃N₄NaO₆S₂·H₂O MW: 492,47 g/mol (anhy) |
MDL Number:
MFCD00150794 CAS Number: 69898-45-9 EINECS: 274-196-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...