3-(4-Piperidinyloxy)benzonitrile hydrochloride ≥97%

Supplier: Thermo Scientific
Warning

H51143.MDEA 65.5 EUR
H51143.MD H51143.03
3-(4-Piperidinyloxy)benzonitrile hydrochloride ≥97%
3-(4-Piperidinyloxy)benzonitrile hydrochloride
Formula: C₁₂H₁₅ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD11977714
CAS Number: 950649-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR