1-(4-Methoxyphenylsulphonyl)pyrrolidine ≥97%

Supplier: Thermo Scientific
Warning

H56035.MDEA 86.4 EUR
H56035.MD H56035.03
1-(4-Methoxyphenylsulphonyl)pyrrolidine ≥97%
1-(4-Methoxyphenylsulphonyl)pyrrolidine
Formula: C₁₁H₁₅NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00783727
CAS Number: 335215-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR