1-(3-Methoxyphenyl)piperazine dihydrochloride
Supplier: Molekula
MOLE17665229-25GEA
0
EUR
MOLE17665229-25G
1-(3-Methoxyphenyl)piperazine dihydrochloride
1-(3-Methoxyphenyl)piperazine dihydrochloride
Formula:
C₁₁H₁₈Cl₂N₂O MW: 265,18 g/mol |
MDL Number:
MFCD00035285 CAS Number: 6968-76-9 EINECS: 230-189-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...