6-Chloro-5-fluorobenzimidazole ≥95%

Supplier: Thermo Scientific

A10904.06EA 0 EUR
A10904.06
6-Chloro-5-fluorobenzimidazole ≥95%
6-Chloro-5-fluorobenzimidazole
Formula: C₇H₄ClFN₂
MW: 170,56 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00156132
CAS Number: 175135-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR