N-Boc-D-valine

Supplier: Novabiochem (Part of Merck)

Synonyms: Boc-D-Val-OH

04-13-0031-25EA 220 EUR
04-13-0031-25
N-Boc-D-valine
N-Boc-D-valine
Formula: C₁₀H₁₉NO₄
MW: 217,3 g/mol
MDL Number: MFCD00038282
CAS Number: 22838-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR