2-Chloro-3,4-dimethoxybenzyl alcohol ≥97%
Supplier: Thermo Scientific
Synonyms:
(2-Chloro-3,4-dimethoxyphenyl)methanol
B20688.06EA
125
EUR
B20688.06
B20688.03
2-Chloro-3,4-dimethoxybenzyl alcohol ≥97%
2-Chloro-3,4-dimethoxybenzyl alcohol
Formula:
C₉H₁₁ClO₃ MW: 202,64 g/mol Melting Pt: 70…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210141 CAS Number: 93983-13-2 EINECS: 301-301-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...