4-Cyanophenyl-4-propylbenzoate ≥99%

Supplier: Thermo Scientific
Danger

B22274.06EA 157 EUR
B22274.06 B22274.03
4-Cyanophenyl-4-propylbenzoate ≥99%
4-Cyanophenyl 4-propylbenzoate
Formula: C₁₇H₁₅NO₂
Melting Pt: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD03410282
CAS Number: 56131-49-8
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR