5-Acetylsalicylamide ≥98%

Supplier: Thermo Scientific
Warning

A18303.06EA 20.2 EUR
A18303.06 A18303.22 A18303.14
5-Acetylsalicylamide ≥98%
5-Acetylsalicylamide
Formula: C₉H₉NO₃
MW: 179,18
Melting Pt: 220…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00049222
CAS Number: 40187-51-7
EINECS: 254-830-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR