4-Chloro-2,5-dimethylbenzenesulphonamide ≥97%

Supplier: Thermo Scientific
Warning

B21683.06EA 229 EUR
B21683.06 B21683.03
4-Chloro-2,5-dimethylbenzenesulphonamide ≥97%
4-Chloro-2,5-dimethylbenzenesulphonamide
Formula: C₈H₁₀ClNO₂S
Melting Pt: 190…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00275263
CAS Number: 219689-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR