Picolinafen, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

37912-100MG-REA 65.3 EUR
37912-100MG-R
Picolinafen, Supelco®
Picolinafen
Formula: C₁₉H₁₂F₄N₂O₂
MDL Number: MFCD04040061
CAS Number: 137641-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR