3-Nitro-4-picoline

Supplier: Molekula

Synonyms: 4-Methyl-3-nitropyridine

MOLE36067304-5GEA 0 EUR
MOLE36067304-5G
3-Nitro-4-picoline
3-Nitro-4-picoline
Formula: C₆H₆N₂O₂
MW: 138,13 g/mol
MDL Number: MFCD00051829
CAS Number: 5832-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR