4-Ethylcaprylic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Ethyloctanoic acid

B22220.88EA 27.7 EUR
B22220.88 B22220.09
4-Ethylcaprylic acid ≥97%
4-Ethylcaprylic acid
Formula: C₁₀H₂₀O₂
MW: 172,27
Boiling Pt: 142…143 °C (10 mmHg)
Density: 0,908
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00506494
CAS Number: 16493-80-4
EINECS: 240-560-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR