3-Amino-2,6-dimethylpyridine 97%

Supplier: Thermo Scientific
Danger

Synonyms: 2,6-Dimethylpyridin-3-amine

L19842.03EA 36.4 EUR
L19842.03 L19842.14 L19842.06
3-Amino-2,6-dimethylpyridine 97%
3-Amino-2,6-dimethylpyridine
Formula: C₇H₁₀N₂
MW: 122,17
Boiling Pt: 230 °C
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD01646409
CAS Number: 3430-33-9
EINECS: 222-332-7
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR