Methyl-2-(methylthio)butyrate ≥98%

Supplier: Thermo Scientific
Warning

B22303.06EA 32.2 EUR
B22303.06 B22303.14
Methyl-2-(methylthio)butyrate ≥98%
Methyl 2-(methylthio)butyrate
Formula: C₆H₁₂O₂S
Boiling Pt: 103…104 °C (35 mmHg)
Density: 1,028
Flash Pt: 64 °C (147 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013393
CAS Number: 51534-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR