Lead(II) trifluoroacetate hemihydrate
Supplier: Thermo Scientific
|
Danger
|
17553.09EA
147
EUR
17553.09
Lead(II) trifluoroacetate hemihydrate
Lead (II) trifluoroacetate
It is employed in pyrolytic deposition of fluoride glass.
Caution: Store in a cool dry place in tightly closed containers. Incompatible with oxidizing agents. Air and moisture sensitive.
Formula:
C₄F₆O₄Pb·0.5H₂O Melting Pt: 147…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061594 CAS Number: 4146-73-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...