1,4-Diisopropylcyclohexane (mixture of cis and trans isomeres) ≥98%

Supplier: Thermo Scientific
Danger

B21587.06EA 56.4 EUR
B21587.06
1,4-Diisopropylcyclohexane (mixture of cis and trans isomeres) ≥98%
1,4-Diisopropylcyclohexane (mixture of cis- and trans-isomeres)
Formula: C₁₂H₂₄
MW: 168,32 g/mol
Boiling Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00019366
CAS Number: 22907-72-8
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR