6-Nitropicolinic acid ≥97%
|
Warning
|
Synonyms:
6-Nitropyridine-2-carboxylic acid
H35766.03EA
319
EUR
H35766.03
H35766.MD
6-Nitropicolinic acid ≥97%
6-Nitropicolinic acid
Formula:
C₆H₄N₂O₄ MW: 168,11 g/mol Melting Pt: 170…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114205 CAS Number: 26893-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...